(5-Chloro-2-methoxy-phenyl)-(4-chlorophenyl)methanone 95%
Leverantör: Apollo Scientific
OR480844-1G OR480844-5G
APOSOR480844-1GEA 1425 SEK
APOSOR480844-1G APOSOR480844-5G
(5-Chloro-2-methoxy-phenyl)-(4-chlorophenyl)methanone 95%
(5-Chloro-2-methoxy-phenyl)-(4-chlorophenyl)methanone
Formel: C₁₄H₁₀Cl₂O₂
MW: 281,134 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11545103
CAS nummer: 76442-96-1