4-Formyl-2-methoxy-benzonitrile 95%

Leverantör: Apollo Scientific

Synonyms: 4-Formyl-2-methoxybenzonitrile, 4-Cyano-3-methoxy-benzaldehyde

OR480828-1G OR480828-5G
APOSOR480828-1GEA 2275 SEK
APOSOR480828-1G APOSOR480828-5G
4-Formyl-2-methoxy-benzonitrile 95%
4-Formyl-2-methoxy-benzonitrile
Formel: C₉H₇NO₂
MW: 161,157 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16295254
CAS nummer: 21962-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR