2,5-Diaminothiophene-3,4-dicarbonitrile 95%

Leverantör: Apollo Scientific

OR480775-1G OR480775-5G
APOSOR480775-1GEA 2425 SEK
APOSOR480775-1G APOSOR480775-5G
2,5-Diaminothiophene-3,4-dicarbonitrile 95%
2,5-Diaminothiophene-3,4-dicarbonitrile
Formel: C₆H₄N₄S
MW: 164,188 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00512774
CAS nummer: 17989-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR