2-Hydroxy-5-methyl-pyridine-3-carboxylic acid 95%
Leverantör: Apollo Scientific
Synonyms: 5-Methyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid
OR480735-1G OR480735-5G OR480735-500MG
APOSOR480735-1GEA 3875 SEK
APOSOR480735-1G APOSOR480735-5G APOSOR480735-500MG
2-Hydroxy-5-methyl-pyridine-3-carboxylic acid 95%
2-Hydroxy-5-methyl-pyridine-3-carboxylic acid
Formel: C₇H₇NO₃
MW: 153,135 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11227183
CAS nummer: 38076-77-6