4-(3-Bromophenyl)-3-methyl-1H-1,2,4-triazol-5-one
Leverantör: Apollo Scientific
Varning
OR47848-1G OR47848-250MG
APOSOR47848-1GEA 6175 SEK
APOSOR47848-1G APOSOR47848-250MG
4-(3-Bromophenyl)-3-methyl-1H-1,2,4-triazol-5-one
4-(3-Bromophenyl)-3-methyl-1H-1,2,4-triazol-5-one
Formel: C₉H₈BrN₃O
MW: 254,083 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18489694
CAS nummer: 259750-15-7