1-Cyclopropyl-1H-pyrazole-4-carboxylic acid
Leverantör: Apollo Scientific
Varning
OR475184-1G OR475184-250MG OR475184-5G
APOSOR475184-1GEA 6750 SEK
APOSOR475184-1G APOSOR475184-250MG APOSOR475184-5G
1-Cyclopropyl-1H-pyrazole-4-carboxylic acid
1-Cyclopropyl-1H-pyrazole-4-carboxylic acid
Formel: C₇H₈N₂O₂
MW: 152,151 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28405628
CAS nummer: 1622883-44-6