N-[(1S)-1-(4-Bromophenyl)ethyl]cyclopropanamine hydrochloride
Leverantör: Apollo Scientific
OR475069-1G OR475069-250MG
APOSOR475069-1GEA 6750 SEK
APOSOR475069-1G APOSOR475069-250MG
N-[(1S)-1-(4-Bromophenyl)ethyl]cyclopropanamine hydrochloride
N-[(1S)-1-(4-Bromophenyl)ethyl]cyclopropanamine hydrochloride
Formel: C₁₁H₁₄BrN·ClH
MW: 276.601 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD31695673
CAS nummer: 2413937-39-8