2-Amino-3-cyanopyridine-5-boronic acid pinacol ester 95%
Leverantör: Apollo Scientific
OR471722-1G OR471722-250MG
APOSOR471722-1GEA 3775 SEK
APOSOR471722-1G APOSOR471722-250MG
2-Amino-3-cyanopyridine-5-boronic acid pinacol ester 95%
2-Amino-3-cyanopyridine-5-boronic acid pinacol ester
Formel: C₁₂H₁₆BN₃O₂
Storage Temperature: Omgivande
MDL Number: MFCD16995200
CAS nummer: 1246372-66-6