Ethyl 2-Cyanopyrimidine-5-carboxylate
Leverantör: Apollo Scientific
OR471577-1G OR471577-250MG
APOSOR471577-1GEA 10775 SEK
APOSOR471577-1G APOSOR471577-250MG
Ethyl 2-Cyanopyrimidine-5-carboxylate
Ethyl 2-Cyanopyrimidine-5-carboxylate
Formel: C₈H₇N₃O₂
Storage Temperature: Omgivande
MDL Number: MFCD29065915
CAS nummer: 2006277-94-5