2,3,5,6-Tetrahydro-1H-imidazo[1,2-a]imidazole hydrochloride
Leverantör: Apollo Scientific
OR471515-1G OR471515-250MG OR471515-5G
APOSOR471515-1GEA 7075 SEK
APOSOR471515-1G APOSOR471515-250MG APOSOR471515-5G
2,3,5,6-Tetrahydro-1H-imidazo[1,2-a]imidazole hydrochloride
2,3,5,6-Tetrahydro-1H-imidazo[1,2-a]imidazole hydrochloride
Formel: C₅H₉N₃·ClH
MW: 147.606 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD29049238
CAS nummer: 120267-00-7