4-(Allyloxy)-2-chloro-6-methylpyrimidine
Leverantör: Apollo Scientific
Synonyms: 2-Chloro-4-methyl-6-(2-propen-1-yloxy)pyrimidine
OR471466-1G OR471466-5G
APOSOR471466-1GEA 3525 SEK
APOSOR471466-1G APOSOR471466-5G
4-(Allyloxy)-2-chloro-6-methylpyrimidine
4-(Allyloxy)-2-chloro-6-methylpyrimidine
Formel: C₈H₉ClN₂O
MW: 184.623 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16767225
CAS nummer: 1250367-45-3