(R)-2-(1-Pyrrolidinyl)propionic acid
Leverantör: Apollo Scientific
Synonyms: (R)-2-(1-Pyrrolidinyl)propionic Acid
OR471316-1G OR471316-5G
APOSOR471316-1GEA 3600 SEK
APOSOR471316-1G APOSOR471316-5G
(R)-2-(1-Pyrrolidinyl)propionic acid
(R)-2-(1-Pyrrolidinyl)propionic acid
Formel: C₇H₁₃NO₂
MW: 143.184 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28716068
CAS nummer: 1234836-32-8