2,4-Dihydroxy-5-methylthieno[2,3-d]pyrimidine
Leverantör: Apollo Scientific
OR471152-1G OR471152-250MG
APOSOR471152-1GEA 9125 SEK
APOSOR471152-1G APOSOR471152-250MG
2,4-Dihydroxy-5-methylthieno[2,3-d]pyrimidine
2,4-Dihydroxy-5-methylthieno[2,3-d]pyrimidine
Formel: C₇H₆N₂O₂S
MW: 182,2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10000843
CAS nummer: 75860-79-6