5-Ethoxynicotinaldehyde

Leverantör: Apollo Scientific

OR470952-1G OR470952-5G
APOSOR470952-1GEA 5875 SEK
APOSOR470952-1G APOSOR470952-5G
5-Ethoxynicotinaldehyde
5-Ethoxynicotinaldehyde
Formel: C₈H₉NO₂
MW: 151.163 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18254211
CAS nummer: 227939-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR