Ethyl 1-Methyl-4-benzimidazolecarboxylate
Leverantör: Apollo Scientific
OR470582-1G OR470582-250MG OR470582-5G
APOSOR470582-1GEA 4675 SEK
APOSOR470582-1G APOSOR470582-250MG APOSOR470582-5G
Ethyl 1-Methyl-4-benzimidazolecarboxylate
Ethyl 1-Methyl-4-benzimidazolecarboxylate
Formel: C₁₁H₁₂N₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD22682796
CAS nummer: 856840-76-1