Ethyl 3-Bromo-7-methylindole-1-carboxylate
Leverantör: Apollo Scientific
OR470499-5G OR470499-1G
APOSOR470499-5GEA 6000 SEK
APOSOR470499-5G APOSOR470499-1G
Ethyl 3-Bromo-7-methylindole-1-carboxylate
Ethyl 3-Bromo-7-methylindole-1-carboxylate
Formel: C₁₂H₁₂BrNO₂
Storage Temperature: Omgivande
MDL Number: MFCD22205877
CAS nummer: 1375064-43-9