cis-7-Cbz-3,7-diazabicyclo[4.2.0]octane
Leverantör: Apollo Scientific
OR470489-1G OR470489-250MG
APOSOR470489-1GEA 31150 SEK
APOSOR470489-1G APOSOR470489-250MG
cis-7-Cbz-3,7-diazabicyclo[4.2.0]octane
cis-7-Cbz-3,7-diazabicyclo[4.2.0]octane
Formel: C₁₄H₁₈N₂O₂
MW: 246.305 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22205862
CAS nummer: 1932046-84-8