1-(Boc-amino)-3-bromo-1-(4-bromophenyl)propane
Leverantör: Apollo Scientific
OR470358-1G OR470358-5G
APOSOR470358-1GEA 5275 SEK
APOSOR470358-1G APOSOR470358-5G
1-(Boc-amino)-3-bromo-1-(4-bromophenyl)propane
1-(Boc-amino)-3-bromo-1-(4-bromophenyl)propane
Formel: C₁₄H₁₉Br₂NO₂
MW: 393.114 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06410623
CAS nummer: 924817-79-8