2-(Fmoc-amino)-2-(1,1-dioxo-4-tetrahydrothiopyranyl)acetic acid
Leverantör: Apollo Scientific
Varning
OR470340-1G OR470340-250MG
APOSOR470340-1GEA 11875 SEK
APOSOR470340-1G APOSOR470340-250MG
2-(Fmoc-amino)-2-(1,1-dioxo-4-tetrahydrothiopyranyl)acetic acid
2-(Fmoc-amino)-2-(1,1-dioxo-4-tetrahydrothiopyranyl)acetic acid
Formel: C₂₂H₂₃NO₆S
Storage Temperature: Omgivande
MDL Number: MFCD02683149
CAS nummer: 369402-98-2