6-(3,4-Dimethoxyphenyl)-3-pyridinecarbaldehyde
Leverantör: Apollo Scientific
OR470338-1G OR470338-5G
APOSOR470338-1GEA 6950 SEK
APOSOR470338-1G APOSOR470338-5G
6-(3,4-Dimethoxyphenyl)-3-pyridinecarbaldehyde
6-(3,4-Dimethoxyphenyl)-3-pyridinecarbaldehyde
Formel: C₁₄H₁₃NO₃
Storage Temperature: Omgivande
MDL Number: MFCD04115484
CAS nummer: 898796-17-3