5-(2,6-Dimethoxyphenyl)-3-pyridinecarbaldehyde
Leverantör: Apollo Scientific
OR470337-1G OR470337-5G
APOSOR470337-1GEA 6950 SEK
APOSOR470337-1G APOSOR470337-5G
5-(2,6-Dimethoxyphenyl)-3-pyridinecarbaldehyde
5-(2,6-Dimethoxyphenyl)-3-pyridinecarbaldehyde
Formel: C₁₄H₁₃NO₃
MW: 243,258 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06410253
CAS nummer: 887973-87-7