6-Phenoxynicotinonitrile

Leverantör: Apollo Scientific

OR470172-1G OR470172-5G
APOSOR470172-1GEA 1200 SEK
APOSOR470172-1G APOSOR470172-5G
6-Phenoxynicotinonitrile
6-Phenoxynicotinonitrile
Formel: C₁₂H₈N₂O
MW: 196,205 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00205454
CAS nummer: 99902-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR