4-Benzyl-2-homomorpholinecarboxylic acid

Leverantör: Apollo Scientific

OR470022-1G OR470022-250MG
APOSOR470022-1GEA 21450 SEK
APOSOR470022-1G APOSOR470022-250MG
4-Benzyl-2-homomorpholinecarboxylic acid
4-Benzyl-2-homomorpholinecarboxylic acid
Formel: C₁₃H₁₇NO₃
MW: 235,279 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11656814
CAS nummer: 1141669-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR