2-Hydroxybutanenitrile
Leverantör: Apollo Scientific
OR46723-1G
OR46723-250MG
OR46723-500MG
OR46723-2.5G
OR46723-5G
APOSOR46723-1GEA
3600
SEK
APOSOR46723-1G
APOSOR46723-250MG
APOSOR46723-500MG
APOSOR46723-2.5G
APOSOR46723-5G
2-Hydroxybutanenitrile
2-Hydroxybutanenitrile
Formel:
C₄H₇NO MW: 85,104 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00042729 CAS nummer: 4476-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...