Cyclopentanecarboximidamide hydrochloride
Leverantör: Apollo Scientific
OR46719-1G OR46719-250MG OR46719-500MG OR46719-2.5G
APOSOR46719-1GEA 4275 SEK
APOSOR46719-1G APOSOR46719-250MG APOSOR46719-500MG APOSOR46719-2.5G
Cyclopentanecarboximidamide hydrochloride
Cyclopentanecarboximidamide hydrochloride
Formel: C₆H₁₂N₂·ClH
MW: 148.634 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08752286
CAS nummer: 68284-02-6