2-[(Methylamino)methylene]propanedinitrile
Leverantör: Apollo Scientific
OR46633-1G OR46633-250MG OR46633-500MG OR46633-2.5G
APOSOR46633-1GEA 5675 SEK
APOSOR46633-1G APOSOR46633-250MG APOSOR46633-500MG APOSOR46633-2.5G
2-[(Methylamino)methylene]propanedinitrile
2-[(Methylamino)methylene]propanedinitrile
Formel: C₅H₅N₃
MW: 107,113 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD19217238
CAS nummer: 79080-32-3