4-Iodostyrene

Leverantör: Apollo Scientific
Varning

OR46241-1G OR46241-250MG OR46241-5G
APOSOR46241-1GEA 2575 SEK
APOSOR46241-1G APOSOR46241-250MG APOSOR46241-5G
4-Iodostyrene
4-Iodostyrene
Formel: C₈H₇I
Smältpunkt: 40…46 °C
Storage Temperature: Frys
MDL Number: MFCD00045321
CAS nummer: 2351-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR