5-Bromo-7-methylimidazo[1,2-a]pyridine
Leverantör: Apollo Scientific
Varning
OR45229-1G OR45229-250MG
APOSOR45229-1GEA 5825 SEK
APOSOR45229-1G APOSOR45229-250MG
5-Bromo-7-methylimidazo[1,2-a]pyridine
5-Bromo-7-methylimidazo[1,2-a]pyridine
Formel: C₈H₇BrN₂
MW: 211,059 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12756242
CAS nummer: 377779-74-3