1-(4-Cyclopropylphenyl)sulphonylpiperazine
Leverantör: Apollo Scientific
Varning
OR45226-1G OR45226-250MG
APOSOR45226-1GEA 8225 SEK
APOSOR45226-1G APOSOR45226-250MG
1-(4-Cyclopropylphenyl)sulphonylpiperazine
1-(4-Cyclopropylphenyl)sulphonylpiperazine
Formel: C₁₃H₁₈N₂O₂S
MW: 266,359 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD32219473
CAS nummer: 2416262-92-3