1-Piperazinepropanamine 3HCl 97%
Leverantör: Apollo Scientific
OR452192-1G
OR452192-250MG
OR452192-500MG
APOSOR452192-1GEA
10625
SEK
APOSOR452192-1G
APOSOR452192-250MG
APOSOR452192-500MG
1-Piperazinepropanamine 3HCl 97%
1-Piperazinepropanamine 3HCl
Formel:
C₇H₁₇N₃·3ClH MW: 252,613 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD11112123 CAS nummer: 52198-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...