(S)-b-Amino-2-chlorobenzenepropanol 97%
Leverantör: Apollo Scientific
OR452174-1G
OR452174-250MG
OR452174-500MG
APOSOR452174-1GEA
10275
SEK
APOSOR452174-1G
APOSOR452174-250MG
APOSOR452174-500MG
(S)-b-Amino-2-chlorobenzenepropanol 97%
(S)-b-Amino-2-chlorobenzenepropanol
Formel:
C₉H₁₂ClNO MW: 185,651 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD17214630 CAS nummer: 103616-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...