1-(2-Phenoxyphenyl)-ethanone 97%
Leverantör: Apollo Scientific
OR452061-1G OR452061-5G
APOSOR452061-1GEA 4025 SEK
APOSOR452061-1G APOSOR452061-5G
1-(2-Phenoxyphenyl)-ethanone 97%
1-(2-Phenoxyphenyl)-ethanone
Formel: C₁₄H₁₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD08271961
CAS nummer: 26388-13-6