4-Chloro-DL-tryptophan 97%
Leverantör: Apollo Scientific
OR452029-1G OR452029-250MG OR452029-500MG
APOSOR452029-1GEA 9225 SEK
APOSOR452029-1G APOSOR452029-250MG APOSOR452029-500MG
4-Chloro-DL-tryptophan 97%
4-Chloro-DL-tryptophan
Formel: C₁₁H₁₁ClN₂O₂
MW: 238.67 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09756722
CAS nummer: 118244-93-2