7-Bromo-D-tryptophan 97%
Leverantör: Apollo Scientific
OR452009-250MG OR452009-500MG
APOSOR452009-250MGEA 8225 SEK
APOSOR452009-250MG APOSOR452009-500MG
7-Bromo-D-tryptophan 97%
7-Bromo-D-tryptophan
Formel: C₁₁H₁₁BrN₂O₂
MW: 283.121 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09264336
CAS nummer: 496929-99-8