1H-Pyrazolo[3,4-b]pyridin-5-amine hydrochloride
Leverantör: Apollo Scientific
Varning
OR451450-1G OR451450-5G
APOSOR451450-1GEA 825 SEK
APOSOR451450-1G APOSOR451450-5G
1H-Pyrazolo[3,4-b]pyridin-5-amine hydrochloride
1H-Pyrazolo[3,4-b]pyridin-5-amine hydrochloride
Formel: C₆H₆N₄·ClH
MW: 170,6 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD25371291
CAS nummer: 1431970-26-1