4-Bromo-1H-pyrazol-3-amine hydrobromide
Leverantör: Apollo Scientific
Varning
OR451440-10G OR451440-1G OR451440-5G
APOSOR451440-10GEA 2125 SEK
APOSOR451440-10G APOSOR451440-1G APOSOR451440-5G
4-Bromo-1H-pyrazol-3-amine hydrobromide
4-Bromo-1H-pyrazol-3-amine hydrobromide
Formel: C₃H₄BrN₃·BrH
MW: 242,9 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08456823
CAS nummer: 16461-99-7