Sodium ethanethiolate 90%
Leverantör: Apollo Scientific
|
Fara
|
OR45141-10G
OR45141-50G
APOSOR45141-10GEA
2100
SEK
APOSOR45141-10G
APOSOR45141-50G
Sodium ethanethiolate 90%
Sodium ethanethiolate
Formel:
C₂H₅S·Na MW: 84,116 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00066318 CAS nummer: 811-51-8 UN: 3263 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...