1,4-Dioxaspiro[4.5]decane-8,8-dimethanol
Leverantör: Apollo Scientific
Varning
OR451157-1G OR451157-250MG
APOSOR451157-1GEA 14625 SEK
APOSOR451157-1G APOSOR451157-250MG
1,4-Dioxaspiro[4.5]decane-8,8-dimethanol
1,4-Dioxaspiro[4.5]decane-8,8-dimethanol
Formel: C₁₀H₁₈O₄
MW: 202,248 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD20260014
CAS nummer: 1256546-72-1