3-Bromo-2-iodothiophene 95+%
Leverantör: Apollo Scientific
OR43638-100MG
OR43638-250MG
APOSOR43638-100MGEA
1100
SEK
APOSOR43638-100MG
APOSOR43638-250MG
3-Bromo-2-iodothiophene 95+%
3-Bromo-2-iodothiophene
Formel:
C₄H₂BrIS MW: 288,932 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD11848091 CAS nummer: 60404-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...