1-[(1,3-Dioxolan-2-yl)methyl]piperazine
Leverantör: Apollo Scientific
Varning
Synonyms: 2-[(Piperazin-1-yl)methyl]-1,3-dioxolane, (Piperazin-1-yl)acetaldehyde ethylene acetal
OR4188-1G OR4188-5G
APOSOR4188-1GEA 1475 SEK
APOSOR4188-1G APOSOR4188-5G
1-[(1,3-Dioxolan-2-yl)methyl]piperazine
1-[(1,3-Dioxolan-2-yl)methyl]piperazine
Formel: C₈H₁₆N₂O₂
MW: 172,225 g/mol
Densitet: 1,492 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD02093541
CAS nummer: 682802-92-2