2-(2-Bromoethyl)benzeneboronic acid, pinacol ester 97%
Leverantör: Apollo Scientific
OR4106-1G OR4106-250MG
APOSOR4106-1GEA 9850 SEK
APOSOR4106-1G APOSOR4106-250MG
2-(2-Bromoethyl)benzeneboronic acid, pinacol ester 97%
2-(2-Bromoethyl)benzeneboronic acid, pinacol ester
Formel: C₁₄H₂₀BBrO₂
MW: 311.022 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06659922
CAS nummer: 850567-53-2