Ethyl 7-methyl-2-(thien-2-yl)pyrazolo[1,5-a]-pyrimidine-6-carboxylate
Leverantör: Apollo Scientific
OR4082-1G OR4082-5G
APOSOR4082-1GEA 945 SEK
APOSOR4082-1G APOSOR4082-5G
Ethyl 7-methyl-2-(thien-2-yl)pyrazolo[1,5-a]-pyrimidine-6-carboxylate
Ethyl 7-methyl-2-(thien-2-yl)pyrazolo[1,5-a]-pyrimidine-6-carboxylate
Formel: C₁₄H₁₃N₃O₂S
MW: 287,337 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00067907
CAS nummer: 162286-69-3