1-Bromo-3,5-diethylbenzene 98%
Leverantör: Apollo Scientific
Varning
OR40750-25G OR40750-5G OR40750-1G
APOSOR40750-25GEA 5250 SEK
APOSOR40750-25G APOSOR40750-5G APOSOR40750-1G
1-Bromo-3,5-diethylbenzene 98%
1-Bromo-3,5-diethylbenzene
Formel: C₁₀H₁₃Br
MW: 213,114 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08062420
CAS nummer: 90267-03-1