3-(3-Methyl-1,2,4-oxadiazol-5-yl)piperidine 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
3-Methyl-5-(piperidin-3-yl)-1,2,4-oxadiazole
OR40720-100MG
OR40720-1G
OR40720-500MG
APOSOR40720-100MGEA
2150
SEK
APOSOR40720-100MG
APOSOR40720-1G
APOSOR40720-500MG
3-(3-Methyl-1,2,4-oxadiazol-5-yl)piperidine 98%
3-(3-Methyl-1,2,4-oxadiazol-5-yl)piperidine
Formel:
C₈H₁₃N₃O MW: 167,208 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD07801211 CAS nummer: 902837-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...