2-Bromo-4-iodobenzoic acid
Leverantör: Apollo Scientific
Varning
OR40656-25G OR40656-5G OR40656-10G
APOSOR40656-25GEA 3175 SEK
APOSOR40656-25G APOSOR40656-5G APOSOR40656-10G
2-Bromo-4-iodobenzoic acid
2-Bromo-4-iodobenzoic acid
Formel: C₇H₄BrIO₂
MW: 326,914 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11520409
CAS nummer: 28547-29-7