2-Bromo-6-iodobenzoic acid
Leverantör: Apollo Scientific
Varning
OR40647-1G OR40647-250MG OR40647-5G
APOSOR40647-1GEA 2025 SEK
APOSOR40647-1G APOSOR40647-250MG APOSOR40647-5G
2-Bromo-6-iodobenzoic acid
2-Bromo-6-iodobenzoic acid
Formel: C₇H₄BrIO₂
MW: 326,914 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11036149
CAS nummer: 1022128-96-6