3-Bromo-4-iodophenol 97%
Leverantör: Apollo Scientific
Varning
OR40578-1G OR40578-250MG OR40578-5G
APOSOR40578-1GEA 1975 SEK
APOSOR40578-1G APOSOR40578-250MG APOSOR40578-5G
3-Bromo-4-iodophenol 97%
3-Bromo-4-iodophenol
Formel: C₆H₄BrIO
MW: 298,904 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD17000017
CAS nummer: 1037298-05-7