Methyl 5-methylisoxazole-4-carboxylate

Leverantör: Apollo Scientific

Synonyms: 4-(Methoxycarbonyl)-5-methylisoxazole

OR40304-1G OR40304-100MG OR40304-250MG OR40304-500MG OR40304-2.5G
APOSOR40304-1GEA 3550 SEK
APOSOR40304-1G APOSOR40304-100MG APOSOR40304-250MG APOSOR40304-500MG APOSOR40304-2.5G
Methyl 5-methylisoxazole-4-carboxylate
Methyl 5-methylisoxazole-4-carboxylate
Formel: C₆H₇NO₃
Storage Temperature: Omgivande
MDL Number: MFCD09756501
CAS nummer: 100047-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR