Methyl 2-iodo-4-nitrobenzoate 95%
Leverantör: Apollo Scientific
OR401029-1G OR401029-5G OR401029-10G
APOSOR401029-1GEA 410 SEK
APOSOR401029-1G APOSOR401029-5G APOSOR401029-10G
Methyl 2-iodo-4-nitrobenzoate 95%
Methyl 2-iodo-4-nitrobenzoate
Formel: C₈H₆INO₄
Storage Temperature: Omgivande
MDL Number: MFCD00277928
CAS nummer: 6326-42-7