Methyl 2-bromo-5-iodobenzoate
Leverantör: Apollo Scientific
OR400951-1G OR400951-5G
APOSOR400951-1GEA 1025 SEK
APOSOR400951-1G APOSOR400951-5G
Methyl 2-bromo-5-iodobenzoate
Methyl 2-bromo-5-iodobenzoate
Formel: C₈H₆BrIO₂
MW: 340,941 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01851253
CAS nummer: 717880-58-5